Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnni3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177066.5 | NP_796040.3 | 4 | 390 | ||
GenBank mRNA | AK052882.1 | - | 4 | 303 | ||
AK084817.1 | - | 4 | 336 | |||
AY526095.1 | - | 4 | 390 | |||
AY526096.1 | - | 4 | 390 | |||
BC139368.1 | - | 4 | 323 | 8862390 | ||
BC139394.1 | - | 4 | 323 | 8862416 |