Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Try4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011646.5 | NP_035776.1 | 2 | 86 | ||
GenBank mRNA | AK003064.1 | - | 2 | 90 | ||
AK008660.1 | - | 2 | 87 | |||
AK008667.1 | - | 2 | 87 | |||
BC050120.1 | - | 2 | 67 | |||
BC061135.1 | - | 2 | 86 | 30306436 |
Gene Symbol: Gm10334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103153.1 | NP_001096623.1 | 2 | 105 |
Gene Symbol: Prss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053243.2 | NP_444473.1 | 2 | 86 | ||
GenBank mRNA | BC094921.1 | - | 2 | 75 | 6433981 |
Gene Symbol: Prss3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011645.2 | NP_035775.1 | 2 | 89 | ||
GenBank mRNA | AK131862.1 | - | 2 | 90 | ||
BC098498.1 | - | 2 | 72 | |||
BC141304.1 | - | 2 | 154 | 9055916 |
Gene Symbol: Try5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003405.4 | NP_001003405.1 | 2 | 88 | ||
GenBank mRNA | AB009661.1 | - | 2 | 88 | ||
AK007843.1 | - | 2 | 87 | |||
AK131898.1 | - | 2 | 87 | |||
AK160484.1 | - | 2 | 87 | |||
BC139220.1 | - | 2 | 173 | 8862242 | ||
BC139223.1 | - | 2 | 173 | 8862245 | ||
BY061854.1 | - | 2 | 97 |