Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020485.4 | NP_065231.3 | 10 | 1451 | ||
NM_138616.3 | NP_619522.3 | 7 | 998 | |||
NM_138617.3 | NP_619523.3 | 7 | 1056 | |||
NM_138618.3 | NP_619524.3 | 9 | 1317 | |||
XM_005245957.3 | XP_005246014.1 | 8 | 1762 | |||
XM_011541888.2 | XP_011540190.1 | 11 | 1951 | |||
XM_011541889.2 | XP_011540191.1 | 10 | 1466 | |||
XM_011541890.2 | XP_011540192.1 | 9 | 1832 | |||
XM_011541891.2 | XP_011540193.1 | 11 | 1568 | |||
XM_017002014.1 | XP_016857503.1 | 8 | 1711 | |||
GenBank mRNA | AF056965.1 | - | 10 | 1363 | ||
AF510065.1 | - | 10 | 1407 | |||
AF510066.1 | - | 10 | 1407 | |||
AF510067.1 | - | 10 | 1407 | |||
AF510068.1 | - | 10 | 1407 | |||
BC139905.1 | - | 10 | 1493 | 40147407 | ||
JX420291.1 | - | 10 | 1365 | |||
JX420292.1 | - | 10 | 1365 | |||
JX420293.1 | - | 10 | 1365 | |||
JX420294.1 | - | 10 | 1365 | |||
JX420295.1 | - | 10 | 1365 | |||
JX420296.1 | - | 10 | 1365 | |||
JX420297.1 | - | 10 | 1365 | |||
JX420298.1 | - | 10 | 1365 | |||
JX420299.1 | - | 10 | 1365 | |||
JX420300.1 | - | 10 | 1365 | |||
JX420301.1 | - | 10 | 1365 | |||
JX420302.1 | - | 10 | 1365 | |||
X54534.1 | - | 10 | 1414 | |||
X63095.1 | - | 7 | 978 | |||
X63096.1 | - | 7 | 918 |