Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134257.3 | NP_599018.3 | 16 | 3705 | ||
GenBank mRNA | AF215669.1 | - | 5 | 1411 | ||
AF215670.1 | - | 6 | 1637 | |||
AK004648.1 | - | 10 | 2874 | |||
AK084729.1 | - | 5 | 1480 | |||
AK158257.1 | - | 5 | 1471 | |||
AK159844.1 | - | 1123 | ||||
AK171666.1 | - | 5 | 1394 | |||
AK219746.1 | - | 149 | ||||
BC033449.1 | - | 7 | 2721 | 3599345 |