Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5771 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038997.2 | NP_001034086.1 | 5 | 731 | ||
GenBank mRNA | BC132026.1 | - | 5 | 731 | 40130303 | |
BC132030.1 | - | 5 | 731 | 40130307 |
Gene Symbol: Prss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053243.2 | NP_444473.1 | 5 | 714 | ||
GenBank mRNA | BC094921.1 | - | 5 | 703 | 6433981 |