Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RHD SIRNA Assays ›
Gene Symbol:
RHD
Gene Name:
Rh blood group D antigen
Gene Aliases:
CD240D, DIIIc, RH, RH30, RHCED, RHDVA(TT), RHDel, RHPII, RHXIII, Rh4, RhDCw, RhII, RhK562-II, RhPI
Chromosome Location:
Chr.1: 25272393 - 25330445 on Build GRCh38
UniGene ID:
Hs.449968
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s12018
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RHD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127691.2
NP_001121163.1
4
570
NM_001282867.1
NP_001269796.1
4
566
NM_001282868.1
NP_001269797.1
4
570
NM_001282869.1
NP_001269798.1
4
570
NM_001282870.1
NP_001269799.1
4
570
NM_001282871.1
NP_001269800.1
4
570
NM_001282872.1
NP_001269801.1
4
570
NM_016124.4
NP_057208.2
4
570
XM_017002015.1
XP_016857504.1
4
663
GenBank mRNA
AB018966.1
-
4
508
AB018967.1
-
4
508
AB018968.1
-
4
508
AB018969.1
-
4
508
AB037270.1
-
4
508
AB046420.1
-
4
508
AB049754.1
-
4
508
AF037626.1
-
4
504
AF177938.1
-
4
508
AF187846.1
-
4
508
AF510069.1
-
4
551
AF510070.1
-
4
551
AJ704215.2
-
4
508
AK297151.1
-
4
663
AM183924.3
-
4
508
AY449381.1
-
4
508
AY449382.1
-
4
508
AY449383.1
-
4
508
AY449384.1
-
4
508
AY449385.1
-
4
508
AY751491.1
-
4
508
AY751492.1
-
4
508
AY751493.1
-
4
508
AY751494.1
-
4
508
AY751495.1
-
4
508
AY751496.1
-
4
508
BC139922.1
-
4
558
40148684
DQ451878.1
-
4
527
EU557240.1
-
4
511
EU557241.1
-
4
511
EU661905.1
-
4
448
EU661906.1
-
4
448
FN555129.1
-
1
22
FR745438.1
-
4
508
FR745439.1
-
4
508
FR745440.1
-
4
508
FR745441.1
-
4
508
HE657775.1
-
4
508
JF436967.1
-
4
508
JN635688.1
-
4
508
JN635689.1
-
4
508
JQ065733.1
-
4
508
JQ424879.1
-
4
566
KT240043.1
-
4
508
L08429.1
-
4
508
LC053447.1
-
4
508
LN714468.1
-
4
508
LN824132.1
-
4
508
S57971.1
-
4
508
S70174.1
-
4
508
S73913.1
-
4
508
S78509.1
-
4
508
S82449.1
-
4
508
X63094.1
-
4
566
X63097.1
-
4
524
Back To Top
More Information
Panther Classification:
Molecular Function -
primary active transporter
Gene Ontology Categories:
Function(s)
Process(es)
organic cation transport
nitrogen utilization
ammonium transmembrane transport
ammonium transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-9bfc7:80/100.66.79.6:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline