Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177566.3 | NP_808234.2 | 4, 5 | 1139 | ||
GenBank mRNA | AK051749.1 | - | 1, 2 | 269 | ||
BC089365.1 | - | 4, 5 | 1139 | 6844583 | ||
BC116343.1 | - | 4, 5 | 1115 | 40098168 | ||
BC116344.1 | - | 4, 5 | 1115 | 40098169 |