Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apcdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133237.3 | NP_573500.2 | 4 | 1391 | ||
GenBank mRNA | AK156319.1 | - | 4 | 1392 | ||
AK172004.1 | - | 1395 | ||||
AK192359.1 | - | 1 | 241 | |||
BC141269.1 | - | 5 | 1165 | 9055881 | ||
BC141270.1 | - | 5 | 1211 | 9055882 |