Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taar9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010831.1 | NP_001010831.1 | 1 | 878 | ||
GenBank mRNA | BC139084.1 | - | 1 | 927 | 8862106 | |
BC139085.1 | - | 1 | 926 | 8862107 |
Set Membership: |
G-protein coupled receptor |