Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ear14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017389.2 | NP_059085.2 | 1 | 180 |
Gene Symbol: Ear4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017422.1 | NP_001017422.1 | 1 | 186 |