Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002931.3 | NP_002922.2 | 4 | 528 | ||
GenBank mRNA | AK289355.1 | - | 4 | 537 | ||
AK301010.1 | - | 4 | 537 | |||
AK301378.1 | - | 4 | 471 | |||
BC002922.2 | - | 4 | 523 | 3954313 | ||
BC051866.2 | - | 4 | 521 | 6142438 | ||
BI259576.1 | - | 4 | 537 | |||
BT007352.1 | - | 2 | 247 | |||
Z14000.1 | - | 2 | 322 |