Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027711.1 | NP_081987.1 | 9 | 1108 | ||
GenBank mRNA | AK142813.1 | - | 9 | 1109 | ||
AK147360.1 | - | 9 | 1104 | |||
BC052916.1 | - | 9 | 948 | |||
BC094939.1 | - | 9 | 936 | |||
BC150958.1 | - | 9 | 1107 | 9087869 |