Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027711.1 | NP_081987.1 | 14, 15 | 1834 | ||
GenBank mRNA | AK142813.1 | - | 14, 15 | 1835 | ||
AK147360.1 | - | 14, 15 | 1828 | |||
BC052916.1 | - | 14, 15 | 1674 | |||
BC094939.1 | - | 14, 15 | 1662 |