Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrfap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026242.3 | NP_080518.1 | 2 | 1146 | ||
GenBank mRNA | AB041651.1 | - | 2 | 1126 | ||
AK005313.1 | - | 2 | 1138 | |||
AK009827.1 | - | 2 | 1104 | |||
AK017832.1 | - | 2 | 1128 | |||
AK018675.1 | - | 2 | 1096 | |||
AK020049.1 | - | 2 | 1114 | |||
AK159464.1 | - | 2 | 1135 | |||
AK167112.1 | - | 2 | 1127 | |||
AK170789.1 | - | 2 | 1096 | |||
AK170909.1 | - | 2 | 1124 | |||
AK170984.1 | - | 2 | 1128 | |||
BC010209.1 | - | 2 | 1115 | 3482091 | ||
BC085488.1 | - | 2 | 991 |
Gene Symbol: 1700011I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CF981605.1 | - | 566 |