Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002933.4 | NP_002924.1 | 2 | 266 | ||
NM_198232.2 | NP_937875.1 | 2 | 336 | |||
NM_198234.2 | NP_937877.1 | 3 | 337 | |||
NM_198235.2 | NP_937878.1 | 3 | 349 | |||
GenBank mRNA | AK098375.1 | - | 2 | 316 | ||
AK129871.1 | - | 2 | 257 | |||
AK312100.1 | - | 2 | 237 | |||
AV710596.1 | - | 3 | 307 | |||
BC005324.1 | - | 2 | 266 | 3950230 | ||
BC022882.1 | - | 2 | 240 | 4294153 | ||
BF106121.1 | - | 214 | ||||
BG539567.1 | - | 2 | 237 | |||
BI599530.1 | - | 2 | 238 | |||
BI753463.1 | - | 2 | 336 | |||
BI760598.1 | - | 2 | 224 | |||
BM007311.1 | - | 2 | 209 | |||
BM919747.1 | - | 2 | 221 | |||
BP385178.1 | - | 1 | 28 | |||
CB993343.1 | - | 3 | 346 | |||
CD703706.1 | - | 3 | 337 | |||
CR450318.1 | - | 1 | 160 |