Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146391.2 | NP_666503.2 | 1 | 794 |
Gene Symbol: Olfr1065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146408.2 | NP_666520.2 | 1 | 818 | ||
GenBank mRNA | BC138401.1 | - | 1 | 818 | 8861421 | |
BC138402.1 | - | 1 | 818 | 8861422 |