Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037502.2 | NP_001032591.1 | 2 | 238 | ||
GenBank mRNA | AK131731.1 | - | 2 | 239 | ||
DQ012039.1 | - | 2 | 238 |