Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5868 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024147.2 | NP_001019318.1 | 3 | 1093 | ||
GenBank mRNA | AK084047.1 | - | 3 | 1094 |