Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021133.3 | NP_066956.1 | 5 | 2074 | ||
GenBank mRNA | AK315231.1 | - | 5 | 2025 | ||
BC090934.1 | - | 5 | 2058 | 3073811 | ||
BC114433.1 | - | 4 | 1940 | 40054016 | ||
BC115697.1 | - | 4 | 1940 | 40054014 | ||
CR627369.1 | - | 5 | 2072 |