Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025380.3 | NP_001020551.3 | 2 | 1055 |
Gene Symbol: Adam25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011781.2 | NP_035911.2 | 2 | 1045 | ||
GenBank mRNA | AF167403.1 | - | 1045 | |||
AK016581.1 | - | 2 | 1043 |