Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185009.2 | NP_001171938.1 | 5 | 1022 | ||
NM_001185010.2 | NP_001171939.1 | 4 | 500 | |||
NM_002938.4 | NP_002929.1 | 4 | 500 | |||
GenBank mRNA | AB000468.1 | - | 4 | 449 | ||
AK128038.1 | - | 4 | 1663 | |||
AK307372.1 | - | 4 | 462 | |||
AK309509.1 | - | 4 | 428 | |||
AK312534.1 | - | 4 | 429 | |||
BC031935.1 | - | 5 | 1022 | 4824114 | ||
DA342512.1 | - | 4 | 500 | |||
U95140.1 | - | 4 | 422 |