Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185009.2 | NP_001171938.1 | 7 | 1194 | ||
NM_002938.4 | NP_002929.1 | 6 | 672 | |||
GenBank mRNA | AB000468.1 | - | 6 | 621 | ||
AK128038.1 | - | 6 | 1835 | |||
AK312534.1 | - | 6 | 601 | |||
BC031935.1 | - | 7 | 1194 | 4824114 | ||
U95140.1 | - | 6 | 594 |