Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccpg1os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198789.1 | NP_001185718.1 | 2 | 522 | ||
GenBank mRNA | BC048630.1 | - | 2 | 505 | ||
BC099568.1 | - | 2 | 499 |