Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033500.3 | NP_001028672.2 | 2 | 535 | ||
GenBank mRNA | AK052055.1 | - | 2 | 539 | ||
AK135824.1 | - | 2 | 515 | |||
AK142559.1 | - | 2 | 515 | |||
BB658361.1 | - | 2 | 535 | |||
BC145956.1 | - | 2 | 421 | |||
BC145958.1 | - | 2 | 420 |