Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011739.3 | NP_035869.1 | 2, 3 | 758 | ||
GenBank mRNA | AK008948.1 | - | 3, 4 | 528 | ||
AK012300.1 | - | 2, 3 | 491 | |||
AK012540.1 | - | 3, 4 | 493 | |||
AK145568.1 | - | 3, 4 | 483 | |||
AK146371.1 | - | 3, 4 | 483 | |||
AK150850.1 | - | 3, 4 | 483 | |||
AK151437.1 | - | 3, 4 | 532 | |||
AK151715.1 | - | 3, 4 | 511 | |||
AK151864.1 | - | 3, 4 | 532 | |||
AK159403.1 | - | 3, 4 | 516 | |||
AK159817.1 | - | 3, 4 | 532 | |||
AK163237.1 | - | 3, 4 | 485 | |||
AK167397.1 | - | 3, 4 | 484 | |||
BC003837.1 | - | 2, 3 | 271 | |||
BC080802.1 | - | 3, 4 | 473 | |||
BC085299.1 | - | 3, 4 | 538 | |||
BC090838.1 | - | 2, 3 | 758 | 6403848 | ||
BC106164.1 | - | 2, 3 | 460 | 3582392 | ||
BC110680.1 | - | 2, 3 | 467 | |||
BC132252.1 | - | 2, 3 | 527 | 40130529 | ||
BC132530.1 | - | 2, 3 | 527 | 40130807 | ||
D87662.1 | - | 2, 3 | 450 | |||
U56243.1 | - | 2, 3 | 433 | |||
U57312.1 | - | 2, 3 | 413 |