Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005977.3 | NP_005968.1 | 5 | 907 | ||
NM_183043.2 | NP_898864.1 | 5 | 648 | |||
NM_183044.2 | NP_898865.1 | 5 | 782 | |||
XM_005266485.2 | XP_005266542.1 | 5 | 1253 | |||
XM_005266486.2 | XP_005266543.1 | 5 | 701 | |||
XM_011535177.2 | XP_011533479.1 | 5 | 1275 | |||
GenBank mRNA | AJ010346.1 | - | 5 | 797 | ||
AJ010347.1 | - | 5 | 598 | |||
AK312435.1 | - | 5 | 852 | |||
AY009109.1 | - | 5 | 730 | |||
BC034688.2 | - | 4 | 548 | 4508997 | ||
CR749238.1 | - | 5 | 839 | |||
KU178309.1 | - | 3 | 437 |