Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085534.2 | NP_001079003.1 | 2, 3 | 236 | ||
GenBank mRNA | CK615904.1 | - | 2, 3 | 207 | ||
CK616445.1 | - | 2, 3 | 236 |