Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085534.2 | NP_001079003.1 | 7 | 610 | ||
GenBank mRNA | CK615904.1 | - | 7 | 581 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK615885.1 | - | 135 | Chr. - - [-] on Build |