Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn34b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081671.1 | NP_001075140.1 | 3 | 514 | ||
GenBank mRNA | AK006680.1 | - | 2 | 472 | ||
BC100353.1 | - | 3 | 514 | 30933439 |