Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145845.1 | NP_001139317.1 | 18 | 3690 | ||
NM_002941.3 | NP_002932.1 | 19 | 3008 | |||
NM_133631.3 | NP_598334.2 | 18 | 3690 | |||
XM_006713277.2 | XP_006713340.1 | 18 | 3659 | |||
XM_011533976.1 | XP_011532278.1 | 20 | 3106 | |||
XM_011533977.2 | XP_011532279.1 | 20 | 2959 | |||
XM_011533978.1 | XP_011532280.1 | 20 | 3106 | |||
XM_011533979.1 | XP_011532281.1 | 20 | 3106 | |||
XM_011533980.1 | XP_011532282.1 | 20 | 3106 | |||
XM_017006982.1 | XP_016862471.1 | 19 | 3018 | |||
XM_017006983.1 | XP_016862472.1 | 17 | 3650 | |||
XM_017006984.1 | XP_016862473.1 | 19 | 3097 | |||
XM_017006985.1 | XP_016862474.1 | 18 | 2759 | |||
GenBank mRNA | AF040990.1 | - | 18 | 2800 | ||
AK299920.1 | - | 9 | 1697 | |||
BC115020.1 | - | 18 | 2791 | 7961997 | ||
BC115022.1 | - | 18 | 2791 | 7962000 | ||
BX538319.1 | - | 11 | 1719 |