Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm14484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025260.2 | NP_001020431.1 | 1 | 153 | ||
GenBank mRNA | BC049545.1 | - | 153 | 6770892 |
Gene Symbol: 1700012L04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029588.3 | NP_083864.1 | 1 | 191 | ||
GenBank mRNA | AK005922.1 | - | 1 | 193 |
Gene Symbol: Gm14474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242947.1 | NP_001229876.1 | 1 | 153 |
Gene Symbol: Gm14475 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242954.1 | NP_001229883.1 | 96 |
Gene Symbol: Gm14476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242950.1 | NP_001229879.1 | 96 |
Gene Symbol: Gm14477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242949.1 | NP_001229878.1 | 96 |
Gene Symbol: Gm14478 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242953.1 | NP_001229882.1 | 135 |
Gene Symbol: Gm14479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242951.1 | NP_001229880.1 | 96 |
Gene Symbol: Gm14482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242952.1 | NP_001229881.1 | 1 | 165 |
Gene Symbol: Gm14483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111037.1 | NP_001104507.1 | 1 | 160 | ||
GenBank mRNA | BC087913.1 | - | 1 | 160 | 6705184 |
Gene Symbol: Gm14501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085537.2 | NP_001079006.1 | 1 | 193 |