Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166691.1 | NP_001160163.1 | 15 | 1824 | ||
NM_003318.4 | NP_003309.2 | 15 | 1815 | |||
XM_011536099.2 | XP_011534401.1 | 15 | 1789 | |||
XM_011536100.2 | XP_011534402.1 | 15 | 1812 | |||
XM_017011242.1 | XP_016866731.1 | 16 | 2344 | |||
GenBank mRNA | AK095599.1 | - | 15 | 2284 | ||
AK292460.1 | - | 15 | 1787 | |||
AK315696.1 | - | 15 | 1732 | |||
AX775733.1 | - | 15 | 1787 | |||
BC000633.2 | - | 15 | 1778 | 3343925 | ||
BC032858.2 | - | 15 | 1780 | 5270012 | ||
X70500.1 | - | 2 | 237 |