Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039530.3 | NP_001034619.2 | 8 | 3429 | ||
GenBank mRNA | AK089685.1 | - | 9 | 3520 | ||
AK220263.1 | - | 3 | 1021 | |||
BC047386.1 | - | 3 | 530 | |||
BC138874.1 | - | 8 | 3544 | 8861896 | ||
BC145452.1 | - | 8 | 3544 | |||
DQ372930.1 | - | 8 | 3593 |