Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trav14d-3-dv8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF479020.1 | - | 2 | 135 | ||
BC147846.1 | - | 2 | 231 | 9087563 | ||
BC147852.1 | - | 2 | 231 | 9087569 | ||
L21700.1 | - | 1 | 54 | |||
U07878.1 | - | 158 | ||||
X76971.1 | - | 1 | 54 |
Gene Symbol: Trav14n-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039793.1 | - | 190 |