Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trav14d-3-dv8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF126457.1 | - | 1 | 164 | ||
AF479020.1 | - | 2 | 245 | |||
BC147846.1 | - | 2 | 341 | 9087563 | ||
BC147852.1 | - | 2 | 341 | 9087569 | ||
L21700.1 | - | 1 | 164 | |||
U07878.1 | - | 268 | ||||
X76971.1 | - | 1 | 164 |
Gene Symbol: Trav14-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC147823.1 | - | 2 | 283 | ||
BC147827.1 | - | 2 | 283 |
Gene Symbol: Trav14n-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039793.1 | - | 300 |