Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trav14d-3-dv8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC147846.1 | - | 2 | 317 | 9087563 | |
BC147852.1 | - | 2 | 317 | 9087569 |