Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025351.1 | NP_001020522.1 | 1, 2 | 66 | ||
GenBank mRNA | AY621335.1 | - | 1, 2 | 66 | ||
DQ012053.1 | - | 1, 2 | 66 |