Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029929.2 | NP_001025100.1 | 11, 12 | 2293 | ||
GenBank mRNA | AK029988.1 | - | 11, 12 | 2294 | ||
AK128979.1 | - | 10, 11 | 2600 | |||
AK171931.1 | - | 11, 12 | 2142 | |||
BC038060.1 | - | 8, 9 | 903 | |||
BC151086.1 | - | 11, 12 | 1965 | 9087999 |