Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029929.2 | NP_001025100.1 | 5, 6 | 1613 | ||
GenBank mRNA | AK029988.1 | - | 5, 6 | 1614 | ||
AK128979.1 | - | 4, 5 | 1920 | |||
AK171931.1 | - | 5, 6 | 1462 | |||
BC038060.1 | - | 2, 3 | 223 | |||
BC151086.1 | - | 5, 6 | 1285 | 9087999 |