Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153160.4 | NP_694800.1 | 4 | 307 | ||
GenBank mRNA | AJ272345.1 | - | 4 | 263 | ||
AK012953.1 | - | 4 | 321 | |||
AK152289.1 | - | 4 | 308 | |||
AK161588.1 | - | 4 | 302 | |||
AK168912.1 | - | 308 | ||||
AY253297.1 | - | 4 | 316 | |||
BC018382.1 | - | 3 | 245 | 4215774 | ||
BC049231.1 | - | 4 | 283 | 4161178 |