Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104581.1 | NP_001098051.1 | 6 | 2023 | ||
GenBank mRNA | AB540948.1 | - | 6 | 2023 |
Gene Symbol: Vmn2r114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102584.1 | NP_001096054.1 | 6 | 2020 | ||
GenBank mRNA | AB540945.1 | - | 6 | 2020 |
Gene Symbol: Vmn2r115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104579.1 | NP_001098049.1 | 6 | 2023 | ||
GenBank mRNA | AB540946.1 | - | 6 | 2023 |
Set Membership: |
G-protein coupled receptor |