Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005353.2 | NP_001005353.1 | 6 | 1163 | ||
NM_013410.3 | NP_037542.1 | 5 | 1046 | |||
NM_203464.2 | NP_982289.1 | 6 | 1052 | |||
XM_017000612.1 | XP_016856101.1 | 5 | 798 | |||
XM_017000613.1 | XP_016856102.1 | 5 | 984 | |||
GenBank mRNA | AK025926.1 | - | 5 | 1010 | ||
AK309057.1 | - | 5 | 754 | |||
BC016180.1 | - | 5 | 946 | 4637988 | ||
BC040224.1 | - | 5 | 922 | 5188877 | ||
BC066944.1 | - | 6 | 1163 | 4819382 | ||
BC146653.1 | - | 6 | 1037 | 40147793 | ||
X60673.1 | - | 5 | 988 |