Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205226.1 | NP_001192155.1 | 48 | 7218 | ||
NM_178078.2 | NP_835179.1 | 48 | 7224 | |||
GenBank mRNA | AK031357.1 | - | 10 | 1480 | ||
AK048177.1 | - | 8 | 2681 | |||
AK079464.1 | - | 9 | 1089 | |||
AK129258.1 | - | 4961 | ||||
BC018281.1 | - | 21 | 3195 | |||
BC027147.1 | - | 12 | 1748 | |||
BC058249.1 | - | 47 | 7034 | |||
BC094620.1 | - | 4 | 374 | |||
BC157948.1 | - | 48 | 7137 | 9088123 | ||
BC158073.1 | - | 48 | 7143 | 9007370 |