Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286076.1 | NP_001273005.1 | 2 | 453 | ||
NM_001297558.1 | NP_001284487.1 | 2 | 260 | |||
NM_002946.4 | NP_002937.1 | 2 | 453 | |||
GenBank mRNA | AK298759.1 | - | 2 | 231 | ||
AK300691.1 | - | 2 | 195 | |||
AK312956.1 | - | 2 | 217 | |||
BC001630.1 | - | 2 | 190 | 3538351 | ||
BC012157.1 | - | 2 | 242 | 4563651 | ||
BC021257.2 | - | 2 | 381 | 4111194 | ||
BX333932.2 | - | 2 | 182 | |||
CR450348.1 | - | 1 | 99 | |||
DB025488.1 | - | 2 | 453 | |||
J05249.1 | - | 2 | 176 |