Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278282.1 | NP_001265211.1 | 7 | 736 | ||
NM_001278283.1 | NP_001265212.1 | 8 | 816 | |||
NM_001278285.1 | NP_001265214.1 | 5 | 644 | |||
NM_001278286.1 | NP_001265215.1 | 6 | 660 | |||
NM_001278288.1 | NP_001265217.1 | 5 | 651 | |||
NM_001278289.1 | NP_001265218.1 | 7 | 800 | |||
NM_001318926.1 | NP_001305855.1 | 7 | 785 | |||
NM_001318927.1 | NP_001305856.1 | 7 | 829 | |||
NM_001318928.1 | NP_001305857.1 | 7 | 680 | |||
NM_001318929.1 | NP_001305858.1 | 6 | 727 | |||
NM_001318930.1 | NP_001305859.1 | 6 | 720 | |||
NM_001318931.1 | NP_001305860.1 | 6 | 682 | |||
NM_006916.2 | NP_008847.1 | 6 | 705 | |||
NM_199229.2 | NP_954699.1 | 6 | 731 | |||
XM_006712677.3 | XP_006712740.1 | 7 | 678 | |||
GenBank mRNA | AJ224326.1 | - | 5 | 473 | ||
AK056028.1 | - | 6 | 642 | |||
AK093658.1 | - | 8 | 747 | |||
AK096199.1 | - | 7 | 567 | |||
AK291035.1 | - | 6 | 658 | |||
AK303184.1 | - | 5 | 571 | |||
AK308741.1 | - | 6 | 569 | |||
AL833120.1 | - | 6 | 654 | |||
BC005148.2 | - | 6 | 639 | 3505010 | ||
BC016764.1 | - | 6 | 668 | 4107315 | ||
BC072401.1 | - | 6 | 640 | 6047438 | ||
BX403091.2 | - | 7 | 691 | |||
BX643719.1 | - | 7 | 755 | |||
CR992340.1 | - | 7 | 701 | |||
DA237285.1 | - | 5 | 578 |