Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278282.1 | NP_001265211.1 | 7 | 740 | ||
NM_001278283.1 | NP_001265212.1 | 8 | 820 | |||
NM_001278285.1 | NP_001265214.1 | 5 | 648 | |||
NM_001278286.1 | NP_001265215.1 | 6 | 664 | |||
NM_001278288.1 | NP_001265217.1 | 5 | 655 | |||
NM_001278289.1 | NP_001265218.1 | 7 | 804 | |||
NM_001318926.1 | NP_001305855.1 | 7 | 789 | |||
NM_001318927.1 | NP_001305856.1 | 7 | 833 | |||
NM_001318928.1 | NP_001305857.1 | 7 | 684 | |||
NM_001318929.1 | NP_001305858.1 | 6 | 731 | |||
NM_001318930.1 | NP_001305859.1 | 6 | 724 | |||
NM_001318931.1 | NP_001305860.1 | 6 | 686 | |||
NM_006916.2 | NP_008847.1 | 6 | 709 | |||
NM_199229.2 | NP_954699.1 | 6 | 735 | |||
XM_006712677.3 | XP_006712740.1 | 7 | 682 | |||
GenBank mRNA | AJ224326.1 | - | 5 | 477 | ||
AK056028.1 | - | 6 | 646 | |||
AK093658.1 | - | 8 | 751 | |||
AK096199.1 | - | 7 | 571 | |||
AK291035.1 | - | 6 | 662 | |||
AK303184.1 | - | 5 | 575 | |||
AK308741.1 | - | 6 | 573 | |||
AL833120.1 | - | 6 | 658 | |||
BC005148.2 | - | 6 | 643 | 3505010 | ||
BC016764.1 | - | 6 | 672 | 4107315 | ||
BC072401.1 | - | 6 | 644 | 6047438 | ||
BX403091.2 | - | 7 | 695 | |||
BX643719.1 | - | 7 | 759 | |||
CR992340.1 | - | 7 | 705 |