Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000967.3 | NP_000958.1 | 3, 4 | 441 | ||
NM_001033853.1 | NP_001029025.1 | 3, 4 | 441 | |||
GenBank mRNA | AB062291.1 | - | 2, 3 | 378 | ||
AB266619.1 | - | 2, 3 | 217 | |||
AK092695.1 | - | 3, 4 | 399 | |||
AK315693.1 | - | 3, 4 | 399 | |||
BC002408.2 | - | 2, 3 | 376 | 2961541 | ||
BC004323.1 | - | 1, 2 | 40 | |||
BC006483.1 | - | 3, 4 | 386 | |||
BC008003.1 | - | 2, 3 | 378 | 3507481 | ||
BC008492.1 | - | 3, 4 | 399 | 4251511 | ||
BC012146.1 | - | 3, 4 | 386 | 4549682 | ||
BC012786.2 | - | 3, 4 | 386 | 3951917 | ||
BC013674.1 | - | 2, 3 | 382 | 4152521 | ||
BC013861.1 | - | 2, 3 | 382 | |||
BC014017.2 | - | 2, 3 | 381 | 4128023 | ||
BC015032.1 | - | 2, 3 | 382 | |||
BC015767.1 | - | 2, 3 | 382 | 4861546 | ||
BC022790.1 | - | 2, 3 | 287 | |||
BC033296.1 | - | 2, 3 | 379 | |||
BC063662.1 | - | 2, 3 | 386 | 3852576 | ||
BC088373.1 | - | 3, 4 | 396 | 6195715 | ||
BC107711.1 | - | 3, 4 | 403 | 6699816 | ||
BC127794.1 | - | 1, 2 | 195 | |||
CR456566.1 | - | 2, 3 | 385 | |||
L22453.1 | - | 2, 3 | 376 | |||
M90054.1 | - | 2, 3 | 359 | |||
X73460.1 | - | 2, 3 | 379 |