Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105190.1 | NP_001098660.1 | 2 | 423 |
Gene Symbol: Vmn2r78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105189.1 | NP_001098659.1 | 2 | 423 |
Set Membership: |
G-protein coupled receptor |