Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000968.3 | NP_000959.2 | 2 | 171 | ||
GenBank mRNA | AB208977.1 | - | 2 | 148 | ||
AK125772.1 | - | 3 | 602 | |||
AK130784.1 | - | 2 | 162 | |||
AK223057.1 | - | 2 | 162 | |||
AK291117.1 | - | 2 | 180 | |||
AK291859.1 | - | 2 | 162 | |||
AK294113.1 | - | 2 | 162 | |||
AK297484.1 | - | 1 | 161 | |||
BC001365.2 | - | 2 | 152 | 3051487 | ||
BC005817.2 | - | 1 | 139 | 3688641 | ||
BC007748.2 | - | 1 | 193 | 4098568 | ||
BC007996.1 | - | 1 | 145 | 3546003 | ||
BC009888.2 | - | 2 | 139 | 4025800 | ||
BC010151.2 | - | 2 | 144 | 3353592 | ||
BC014653.1 | - | 2 | 140 | 4636179 | ||
BC018840.2 | - | 2 | 152 | |||
BC040610.1 | - | 1 | 147 | |||
BC066925.1 | - | 2 | 162 | 5259747 | ||
BC095427.1 | - | 2 | 155 | 30711782 | ||
CB149679.1 | - | 2 | 171 | |||
D23660.1 | - | 2 | 162 |