Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm11780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277919.1 | NP_001264848.1 | 1 | 903 | ||
GenBank mRNA | BU743706.1 | - | 1 | 386 |